Order Entry
Austria
ContactUsLinkComponent
Perchloric acid 69 - 72%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Perchloric acid 69 - 72%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Catalog # 9653-00
Supplier:  Avantor
CAS Number:  
undefined
Perchloric acid 69 - 72%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Catalog # 9653-00
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, poly-coated
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    69.0 - 72.0 %
  • ACS - Assay (HClO₄):
    69.0 - 72.0 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Ignition:
    ≤10 ppm
  • Chloride (Cl):
    ≤10 ppm
  • Nitrogen Compounds (as N):
    ≤10 ppm
  • ACS - Silicate and Phosphate (as SiO₂):
    ≤5 ppm
  • Sulfate (SO₄):
    ≤10 ppm
  • Trace Impurities - Aluminum (Al):
    ≤0.050 ppm
  • Trace Impurities - Barium (Ba):
    ≤0.100 ppm
  • Trace Impurities - Cadmium (Cd):
    ≤0.005 ppm
  • Trace Impurities - Calcium (Ca):
    ≤1 ppm
  • Trace Impurities - Chromium (Cr):
    ≤0.050 ppm
  • Trace Impurities - Cobalt (Co):
    ≤0.005 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.050 ppm
  • Heavy Metals (as Pb):
    ≤0.1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.050 ppm
  • Trace Impurities - Lead (Pb):
    ≤0.010 ppm
  • Trace Impurities - Lithium (Li):
    ≤0.100 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤0.050 ppm
  • Trace Impurities - Manganese (Mn):
    ≤0.005 ppm
  • Trace Impurities - Mercury (Hg):
    ≤0.005 ppm
  • Trace Impurities - Nickel (Ni):
    ≤0.005 ppm
  • Trace Impurities - Potassium (K):
    ≤0.500 ppm
  • Trace Impurities - Silicon (Si):
    ≤0.100 ppm
  • Trace Impurities - Silver (Ag):
    ≤0.005 ppm
  • Trace Impurities - Sodium (Na):
    ≤1.000 ppm
  • Trace Impurities - Strontium (Sr):
    ≤0.020 ppm
  • Trace Impurities - Tin (Sn):
    ≤0.010 ppm
  • Trace Impurities - Zinc (Zn):
    ≤0.050 ppm

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
 :GHS03
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325