

Hydrochloric acid 36.5 - 38.0% NF, Multi-Compendial, J.T.Baker®
: Avantor
:7647-01-0
:
:HCl
:MFCD00011324
:110 °C (1013 hPa)
:–30 °C
:1.18 g/cm³ (20 °C)
:Ambient
:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
:231-595-7
:1789
:GHS05
:GHS07
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Pharmaceutical Chemical:= 0.008 %
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- NF - Assay (HCl):36.5 - 38.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Free Chlorine (as Cl₂):≤ 4 ppm
- EP/BP - Sulfate (SO₄):≤ 20 ppm
- EP/BP - Residue on Evaporation:≤ 0.01 %
- EP/BP - Assay (HCl):35.0 - 39.0 %
- JP - Assay (HCl):35.0 - 38.0 %
- JP - Identification:Passes Test
- JP - Residue on ignition, mg/10mL:≤ 1.0
- JP - Bromide or Iodide:Passes Test
- JP - Bromine or Chlorine:Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Sulfite (SO₃):Passes Test
- No Class 1, 2, 3 or other solvents are used or produced in the manufacturing or purification of the product.:= 1 ppm
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:= 5 ppm
- JP - Mercury (Hg):= 0.04 ppm
Frequently Bought Together