Order Entry
Austria
ContactUsLinkComponent
 
 
 
Magnesium sulphate, anhydrous, powder, AR®, Macron Fine Chemicals™
  6070-26
 :  
undefined
 
 
Magnesium sulphate, anhydrous, powder, AR®, Macron Fine Chemicals™
  6070-26
 :  

 

  • Pack type:
    Fibre drum
  • Pk:
    200 lb
  • Seq:
    7197

 

 

  • Ammonium (NH₄):
    ≤0.005 %
  • Arsenic (As):
    ≤0.001 %
  • Calcium (Ca):
    ≤0.04 %
  • Chloride (Cl):
    ≤0.001 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Insoluble Matter:
    ≤0.01 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Loss on Ignition:
    ≤2.0 %
  • Thru U.S. No. 200 Sieve:
    ≥80 %
  • Nitrate (NO₃):
    ≤0.005 %
  • pH of 5% Solution:
    5.0 - 9.2

 

 

 :
 :120.37 g/mol
 :12,05731
 :110 °C (1013 hPa)
 :1127 °C
 :2.65 g/cm³ (20 °C)
 :231-298-2
 :8,II
 :GHS05
 :GHS07
 :Ambient
 :7487-88-9
 :MgSO₄
 :MFCD00011110