Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 30%, TECHNICAL
Corrosive
Irritant
Hydrochloric acid 30%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Hydrochloric acid 30%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    29 - 31 %

Specification Test Results

Frequently Bought Together