

Hydrochloric acid 25%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
:7647-01-0
:
:HCl
:MFCD00011324
:110 °C (1013 hPa)
:–30 °C
:1.18 g/cm³ (20 °C)
:231-595-7
:1789
:8,II
:36.46 g/mol
:GHS05
:GHS07
:Ambient
- Assay:min. 25 %
- Ammonium (NH4):max. 3 ppm
- Free Chlorine:max. 0.5 ppm
- Residue after Ignition:max. 5 ppm
- Sulfate (SO4):max. 1 ppm
- Sulfite (SO3):max. 2 ppm
- Silver (Ag):max. 0.1
- Aluminium (Al):max. 0.5
- Arsenic (As):max. 0.1
- Barium (Ba):max. 0.1
- Beryllium (Be):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.1
- Cobalt (Co):max. 0.1
- Chromium (Cr):max. 0.1
- Copper (Cu):max. 0.1
- Iron (Fe):max. 0.5
- Germanium (Ge):max. 0.1
- Potassium (K):max. 0.1
- Lithium (Li):max. 0.1
- Magnesium (Mg):max. 0.5
- Manganese (Mn):max. 0.1
- Molybdenum (Mo):max. 0.1
- Sodium (Na):max. 0.5
- Nickel (Ni):max. 0.1
- Lead (Pb):max. 0.1
- Strontium (Sr):max. 0.1
- Titanium (Ti):max. 0.1
- Thallium (Tl):max. 0.1
- Vanadium (V):max. 0.1
- Zinc (Zn):max. 0.5
- Zirconium (Zr):max. 0.1
Frequently Bought Together