Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 10% Ph. Eur.
Irritant
Corrosive
Hydrochloric acid 10% Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Hydrochloric acid 10% Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    9.5 - 10.5 %
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Appearance test:
    Clear colourless liquid
  • Free chlorine:
    Max. 1 ppm
  • SO4 (Sulphate):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Residue on evaporation:
    Max. 0.01 %
  • Residual solvents:
    Unlikely by manuf.process

Specification Test Results

Frequently Bought Together