Hydrochloric acid 10% (v/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:∼100 °C (1013 hPa)
Density:1.02…1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together