Order Entry
Austria
ContactUsLinkComponent
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Catalog # HYCLSH30033.01
CAS Number:  
undefined
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Catalog # HYCLSH30033.01
CAS Number:  

Specifications

  • Pack type:
    PETG bottle, square
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Endotoxin concentration:
    ≤1.0 EU/mL
  • Sterilization:
    0.1 µm sterile filtered
  • pH:
    7.9 - 9.4

Specification Test Results

About this item

 :
 :84.01 g/mol
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528