Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium dodecyl sulphate (SDS)
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
A3942.1000
:
PanReac AppliChem
:
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
A3942.1000
:
PanReac AppliChem
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
288.38 g/mol
:
1…1.1 g/cm³ (20 °C)
:
Ambient
:
205-788-1
:
151-21-3
:
CH₃(CH₂)₁₁OSO₃Na
:
MFCD00036175