Order Entry
Austria
ContactUsLinkComponent
Corrosive
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution, Electran for electrophoresis
Catalog # 444062F
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution, Electran for electrophoresis
Catalog # 444062F
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay (W/V) (at preparation):
    9.8 - 10.2 %

Specification Test Results

About this item

 :
 :288.38 g/mol
Density:1…1.1 g/cm³ (20 °C)
EINECS:205-788-1
 :GHS05
Storage Temperature:Ambient
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175