Order Entry
Austria
ContactUsLinkComponent
Trimethylamine 40% (w/w) in aqueous solution for synthesis, Sigma-Aldrich®
Trimethylamine 40% (w/w) in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Trimethylamine 40% (w/w) in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -2 °C
  • Boiling Point:
    30 - 100 °C
  • Vapor Pressure:
    600 hPa (20 °C)
  • Flash Point:
    -45 °C
  • Explosion Limit:
    2 - 11.6 %(V) (anhydrous substance)
  • Density:
    0.88 g/cm3 (20 °C)
  • pH:
    13 (H2O, 20 °C)
  • Ignition Point:
    250 °C

Specification Test Results

Related Information

Frequently Bought Together