Order Entry
Austria
ContactUsLinkComponent
Formaldehyde 38% in aqueous solution, J.T.Baker®
Formaldehyde 38% in aqueous solution, J.T.Baker®
Catalog # 3859.5000
Supplier:  Avantor
CAS Number:  
Formaldehyde 38% in aqueous solution, J.T.Baker®
Catalog # 3859.5000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Formaldehyde (HCHO)(after dilution):
    36 - 40 g/l
  • pH 25°C (after dilution):
    6.8 - 7.2
  • Total Phosphate:
    max. 150 mM

Specification Test Results

About this item

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
 :
 :30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:2209
ADR:8,III
 :GHS06
 :GHS05
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274