Formaldehyde 37% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:200-001-8
UN:2209
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together