Formaldehyde 37% in aqueous solution
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
2 Options Available Below
Product Details & Documents
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



