Formaldehyde 35% in aqueous solution stabilised, TECHNICAL
Supplier: VWR Chemicals
About this item
Warning : Store this product at ambient temperature (above 15°C), because it may polymerise at temperatures below 9°C. Presence of polymers does not affect product properties. Precipitate can be removed by filtration.
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.088 g/cm³ (20 °C)
- Flash Pt:66 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
- :Stabilised with methanol 10-15 %
3 Options Available Below
Product Details & Documents
Warning : Store this product at ambient temperature (above 15°C), because it may polymerise at temperatures below 9°C. Presence of polymers does not affect product properties. Precipitate can be removed by filtration.
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.088 g/cm³ (20 °C)
- Flash Pt:66 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
- :Stabilised with methanol 10-15 %
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
Specifications for Product Family
Specification Test Results
- Assay:Min. 34.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



