Orders LinkContactUsLinkComponent
Validation bottles, Nalgene® | Avantor
Validation bottles, Nalgene®
   Sustainable Options Available
Validation bottles, Nalgene®
   Sustainable Options Available
Certificates
Validation bottles, Nalgene®
3 Options Available Below
Product Details & Documents
PC, clear, with PP screw cap.
  • Thick walls
  • Non-toxic
  • Fully autoclavable
Validation bottles provide material compatibility validation when scaling up production to vessels of larger sizes.
Specifications for Product Family
Specifications
  • Catalog No:
  • NALGDS2127-0250
  • NALGDS2127-0030
  • NALGDS2127-2000
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Thread
    Neck I-Ø
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    53B
    45 mm
    135 mm
    74 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 535,00 €
    535,00 €
    1 * 6 ST
       535,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALGDS2127-0250
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 30 ml
    20-415
    14 mm
    75 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 805,00 €
    805,00 €
    1 * 30 ST
       805,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALGDS2127-0030
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    53B
    42 mm
    270 mm
    123 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 744,00 €
    744,00 €
    1 * 12 ST
       744,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALGDS2127-2000
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact