Order Entry
Austria
ContactUsLinkComponent
Methanol-[D₄] (99.95% D) for NMR spectroscopy, Sigma-Aldrich®
Flammable
Toxic
Health
Methanol-[D₄] (99.95% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.06025.0009
Supplier:  Merck
CAS Number:  
Flammable
Toxic
Health
Methanol-[D₄] (99.95% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.06025.0009
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    7,5 mL
  • Seq:
    2260

Specifications

About this item

 :
 :36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3 (6.1),II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS06
 :GHS08
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637