Order Entry
Austria
ContactUsLinkComponent
Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy
Flammable
Irritant
Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy
Catalog # ACRO325410100
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy
Catalog # ACRO325410100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Ampoule
  • Pk:
    10 mL
  • Seq:
    2076
  • Pk Info:
    10 x 1 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.90 %

Specification Test Results

About this item

 :
 :44.03 g/mol
Boiling Pt:80.7 °C (1013 hPa)
Melting Pt:–46 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
 :GHS02
 :GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881