Order Entry
Austria
ContactUsLinkComponent
Xylene (mixture of isomers) ≥99.5% (by GC, sum of isomers) semiconductor grade
Xylene (mixture of isomers) ≥99.5% (by GC, sum of isomers) semiconductor grade
Certificates
undefined
Xylene (mixture of isomers) ≥99.5% (by GC, sum of isomers) semiconductor grade
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless
  • Form:
    Liquid
  • Assay (GC), (Sum of Isomers+Ethylbenzene+Toluene):
    ≥99.5%
  • Chloride content:
    Max. 3 ppm
  • Color (APHA):
    Max. 10
  • Free Acid (Titration):
    Max. 0.0003 meq/g
  • Impurity Content, B:
    Max. 20 ppb
  • Impurity Content, Phosphate:
    Max. 0.5 ppm
  • Impurity Content, Si:
    Max. 200 ppb
  • Water Content (Karl Fischer Titration):
    Max. 0.02%
  • Total Metal Impurities:
    <1 ppm
  • Heavy Metals (as Pb):
    Max. 1000 ppb
  • Particle Size (Particle Count at Point of fill):
    1 micron and greater: Max. 10 particles/ml
  • Residue on Evaporation:
    Max. 5 ppm
  • Trace Metal:
    Max. 100 ppb (each of: Sn, Na, K, Ge, Pb)

Specification Test Results

Related Information

Frequently Bought Together