Bottles, economy, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, amber with amber PP screw cap.
- Reduce UV light transmissions to protect light-sensitive liquids
- Lightweight
- Good chemical and temperature resistance
Conduct light-duty, general purpose applications where a heavier bottle is not required with these bottles.
Specifications
- Catalog No:
- 331-4223
- 331-4226
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement