Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE, Amber, with Screw Caps
Supplier: Thermo Fisher Scientific
Certificates
About this item
Bottles with closure, suitable for storage and shipping of small volume, light sensitive and valuable reagents.
- Non-cytotoxic and leakproof
- Opaque, linerless polypropylene closures
- Durable, uniform walls that resist splitting and puncturing
Bottles provide light resistance with the ability to see liquid level and are ideal for test kits and many small-volume applications.
Certifications: Compliance with ISO 13485:2003; Meets the requirement of 21CFR177.1520 and USP Class VI.
Ordering information: Product certificate providing cytotoxicity, USP Class VI, FDA Compliance-21 CFR and other regulatory compliance information is provided based upon request.
Packaging: Bottles and closures are packed in separate polybags within the same carton.
Specifications
- Catalog No:
- NALG312084-9125
- NALG312004-9050
- NALG312084-9050
- NALG312004-9025
- NALG312004-9125
- NALG312084-9025
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement