Chemical resistant gloves, KLEENGUARD* G80, neoprene/latex
These hand-specific gloves with cotton flock lining provide chemical hand protection against oxidising acids, bases, alcohols, oils, fats and solvents.
- High abrasion and good tear resistance for better hand protection and high durability
- Ergonomic design, providing comfort and reducing hand fatigue
- Flock lined for extra comfort and easy donning
- Textured palm and fingertips provide excellent grip in wet and dry situations
Made of natural latex covered with black neoprene at the palm, they are suitable for use with many common hydrocarbons and therefore ideal for use in the petrochemical industry.
: EN 374 (AKL), EN 388 (1010), EN 374-2 (Level 2), AQL 1.5
Frequently Bought Together