Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
Supplier:   Brand
Separating funnels
Supplier:  Brand
Certificates
Separating funnels
Separating funnels
4 Options Available Below
Product Details & Documents
PP, high clarity, PTFE stopcock, PP screw cap.
  • Leakproof closure and stopcock
  • Excellent chemical resistance
  • Autoclavable except for stopcock assembly
  • PTFE stopcock requires no lubrication
High clarity for observation of contents.
Product Family Options
Product Information
  • Capacity
    Ø cap
    Availability
    Price
  • 125 ml
    28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 163,00 €
    163,00 €
    1 * 1 ST
       163,00 €
  • 250 ml
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 168,00 €
    168,00 €
    1 * 1 ST
       168,00 €
  • 500 ml
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €
  • 1000 ml
    53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 229,00 €
    229,00 €
    1 * 1 ST
       229,00 €