ContactUsLinkComponent
Fibrillar Collagen 24-Well Multiwell Insert System, Corning® BioCoat®
Fibrillar Collagen 24-Well Multiwell Insert System, Corning® BioCoat®
Catalog # 734-1079
Supplier:  Corning
   Low Manufacturing Impact
undefined
Fibrillar Collagen 24-Well Multiwell Insert System, Corning® BioCoat®
Catalog # 734-1079
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Membrane:
    PET
  • Pore size:
    1,0 µm
  • Description:
    Fibrillar collagen 24-multiwell inserts, with feeder tray and lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Membrane Ø:
    6,4 mm
  • Pk:
    1
Specifications
About this item
This system is packaged with ready-to-use Falcon® Companion Plates.