Order Entry
Austria
ContactUsLinkComponent
Adenovirus purification kits, Vivapure® AdenoPACK™
Adenovirus purification kits, Vivapure® AdenoPACK™
Supplier:  Sartorius
Certificates
undefined
Adenovirus purification kits, Vivapure® AdenoPACK™
Supplier:  Sartorius

Specifications

  • Refine target molecule:
    Viral DNA
  • Sample type:
    Cultured cells
  • Target molecule:
    DNA

Specifications

Related Information

Frequently Bought Together