
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7440-66-6
:
Formula:Zn
MDL Number:MFCD00011291
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:906 °C (1013 hPa)
Melting Pt:419 °C
Density:7.14 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:231-175-3
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:419 °C
- Molar Mass:65.39 g/mol
- Boiling Point:906 °C (1013 hPa)
- Density:7.13 g/cm3 (20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberCertipur Merck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Titration goes Digital
Product Literature
msds_102409
SDS
ProductPage_102409
Product specification
Frequently Bought Together