Order Entry
Austria
ContactUsLinkComponent
Copper, Supelco®
Copper, Supelco®
Catalog # 1.02700.0250
Supplier:  Merck
CAS Number:  
Copper, Supelco®
Catalog # 1.02700.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Melting Point:
    1083 °C
  • Molar Mass:
    63.55 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    2580 °C (1013 hPa)
  • Density:
    8.96 g/cm3

Specification Test Results

About this item

 :
 :63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8.94 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-159-6
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965