Order Entry
Austria
Orders LinkContactUsLinkComponent
Copper, rods

Copper, rods

183 cm
Catalog # 45189.AX
Copper, rods
Catalog # 45189.AX
CAS Number:  7440-50-8
Copper, rods

183 cm

CAS Number:  7440-50-8
Price
Inquire For Price
Log in for availability
1 * 183 cm
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Merck Index:13,02545
  • Boiling Pt:2595 °C (1013 hPa)
  • Melting Pt:1083 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6

Documents

Specifications

Specifications

  • Pk:
    183 cm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.