Orders LinkContactUsLinkComponent
Filter funnels
Filter funnels
Supplier:   Brand
Filter funnels
Supplier:  Brand
Certificates
Filter funnels
8 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Product Family Options
Product Information
  • Capacity
    Plate Ø
    Porosity
    Stem O-Ø
    Availability
    Price
  • 50 ml
    35 mm
    3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,70 €
    55,70 €
    1 * 1 ST
       55,70 €
  • 50 ml
    35 mm
    4
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,00 €
    60,00 €
    1 * 1 ST
       60,00 €
  • 75 ml
    45 mm
    3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,60 €
    62,60 €
    1 * 1 ST
       62,60 €
  • 75 ml
    45 mm
    4
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,00 €
    69,00 €
    1 * 1 ST
       69,00 €
  • 125 ml
    60 mm
    3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,60 €
    87,60 €
    1 * 1 ST
       87,60 €
  • 125 ml
    60 mm
    4
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,70 €
    98,70 €
    1 * 1 ST
       98,70 €
  • 500 ml
    95 mm
    3
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 500 ml
    95 mm
    4
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €