Order Entry
Austria
ContactUsLinkComponent
 
Integral vent filters, PolyVENT™, Whatman™
 
undefined
Integral vent filters, PolyVENT™, Whatman™

 

  • Catalog No:
  • 516-4455
  • WHAT6713-1651
  • 516-4456
  • 516-4457
  • 516-4459
  • Ø:
  • 25 mm
  • 50 mm
  • 75 mm
  • 50 mm
  • 36 mm
  • Housing type:
  • PolyVENT™ 4 disc
  • PolyVENT™ 16 disc
  • PolyVENT™ 1000 capsule
  • PolyVENT™ 16 disc
  • PolyVENT™ 500 capsule
  • Max. pressure:
  • 2 bar
  • 2 bar
  • 2 bar
  • 2 bar
  • 2 bar
  • Inlet/outlet:
  • Female Luer lock/male Luer
  • 1/4"-3/8" stepped barb for tubing Ø 6 - 10 mm
  • 3/8"-1/2" stepped barb for tubing Ø 10 - 13 mm
  • 1/4"-3/8" stepped barb for tubing Ø 6 - 10 mm
  • 1/4"-3/8" stepped barb for tubing Ø 6 - 10 mm

 

Frequently Bought Together