Storage racks for cryoboxes
: GLW
Stainless steel.
- For 130×130 mm cryoboxes
- With handle
Fit any freezer. For easy transport and freezing of samples down to −90 °C.
Frequently Bought Together
Stainless steel.
Fit any freezer. For easy transport and freezing of samples down to −90 °C.