Iodine Reag. Ph. Eur., ACS, ISO analytical reagent, sublimated, Supelco®
: Merck
:7553-56-2
:
:I₂
:MFCD00011355
:253.81 g/mol
:13,05036
:184.3 °C (1013 hPa)
:114 °C
:4.9 g/cm³ (20 °C)
:Ambient
:3495
:8 (6.1),III
:M-Clarity™ quality level = MQ100
:231-442-4
:GHS08
:GHS07
:GHS09
- Environmentally Preferable:
- Solubility:0.3 g/l (20 °C)
- Melting Point:114 °C
- Molar Mass:253.8 g/mol
- Bulk Density:2100 kg/m3
- Boiling Point:185 °C (1013 hPa)
- Vapor Pressure:0.41 hPa (25 °C)
- Density:4.93 g/cm3 (20 °C)
- pH:5.4 (H2O) (saturated solution)
- Viscosity Kinematic:0.57 mm2/s (116 °C) liquid
Frequently Bought Together