


Hydrobromic acid 32% (w/w) in acetic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-113-0
UN:2920
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS07
Specification Test Results
- Bulk Density:(20 °C) soluble
- Boiling Point:- 200 °C (1013 hPa)
- Vapor Pressure:360 hPa (20 °C)
- Density:1.38 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together