Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Selenium, Supelco®

Toxic
Health
Supplier:   Merck
Selenium, Supelco®
Supplier:  Merck
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ300
  • UN:3288
Specifications for Product Family

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    217 °C
  • Molar Mass:
    78.96 g/mol
  • Bulk Density:
    1270 kg/m3
  • Boiling Point:
    685 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 128,00 €
    128,00 €
    1 * 50 g
       128,00 €

  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 854,00 €
    854,00 €
    1 * 250 g
       854,00 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 2 130,00 €
    2 130,00 €
    1 * 1 kg
       2 130,00 €