ContactUsLinkComponent
96-Well T-cell activation plates, Corning® BioCoat™
96-Well T-cell activation plates, Corning® BioCoat™
Catalog # 734-1075
Supplier:  Corning
   Low Manufacturing Impact
undefined
96-Well T-cell activation plates, Corning® BioCoat™
Catalog # 734-1075
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Description:
    T-cell activation 96-well assay plates, anti-human CD3
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Pk:
    5
Specifications
About this item
Corning BioCoat T-Cell activation plates are precoated with high quality BD Pharmingen™ CD3 antibodies. Available for use with mouse or human T-cells, Corning BioCoat T-Cell activation plates offer lot-to-lot consistency and come individually packaged with lids for ease of use.
  • Tested for ability to proliferate mouse splenocytes or human PBMCs
  • Tested and found negative for bacteria and fungi
Applications include:
- T-Cell activation
- Cytokine production
- Cytokine mRNA quantitation
- Co-stimulation
- Studies of drug effects on T-cell function