Funnels with short stem
Supplier: Brand
Certificates
About this item
PP, translucent.
- Excellent chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
With smooth interior surfaces.
Frequently Bought Together
PP, translucent.
With smooth interior surfaces.