Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample cups (PS) swith snap caps (PE)

Sample cups (PS) swith snap caps (PE)

12,00 ml, 24,5 mm, 38 mm, Transparent, White
Catalog # BRND722060
Supplier:   Brand
Sample cups (PS) swith snap caps (PE)
Catalog # BRND722060
Supplier:  Brand
Sample cups (PS) swith snap caps (PE)
Sample cups (PS) swith snap caps (PE)

12,00 ml, 24,5 mm, 38 mm, Transparent, White

Price
171,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Small sample container with white press-on lid.

Specifications
  • Ext.Ø:
    24,5 mm
  • Capacity:
    12,00 ml
  • Colour:
    Transparent
  • Height:
    38 mm
  • Cap colour:
    White
  • Cat. No.:
    BRND722060
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.