Sample cups for Technicon®-Analyser
: Brand
PS, transparent.
PS, transparent.
Product Family Options
- DescriptionCapacityHeightExt.ØPacked
- For Technicon®-Analyser1,5 ml22,7 mm15 mmBag of 10001 * 1.000 ST35,00 €
- For Technicon®-Analyser2,0 ml24,9 mm14,8 mmBag of 10001 * 1.000 ST36,20 €
- For Technicon®-Analyser4,0 ml38 mm17 mmBag of 10001 * 1.000 ST49,50 €
Push-on caps
- DescriptionCapacityHeightExt.ØPacked
- Caps for Technicon 1,5 ml and 2 ml sample cupsBag of 10001 * 1.000 ST35,70 €