Order Entry
Austria
ContactUsLinkComponent
BACC Genomic DNA extraction kits, Nucleon™
BACC Genomic DNA extraction kits, Nucleon™
Catalog # RPN8512
Supplier:  Cytiva
CAS Number:  
undefined
BACC Genomic DNA extraction kits, Nucleon™
Catalog # RPN8512
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Affinity resin
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
    Cultured cells
  • Target molecule:
    DNA
  • Description:
    Nucleon™ BACC3, 50 preps of 10 ml
  • Description:
    Nucleon™ BACC3
  • Yield:
    370-440 µg DNA/mg tissue

Specifications

About this item

Nucleon™ BACC Genomic DNA Extraction Kits were developed for rapid extraction of high quality, high molecular weight genomic DNA from blood and cell cultures.

  • Developed for rapid extraction of high-quality, high molecular weight genomic DNA from whole blood and cell cultures
  • Recovers DNA ranging in size from 23 to 250 kbp as determined by pulse-field gel electrophoresis
  • Features a phenol-free protocol requiring only 30 min to complete

The Nucleon™ proprietary resin is added following cell lysis, deproteinisation with sodium perchlorate, and a single chloroform extraction. Aspiration of the top layer of nucleic acids is simplified and volume of usable sample maximized by a barrier formed between the two layers.

Parameter
Application: Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood.
Protocol length: 30 min
Sample type: Cell cultures and blood
Yield: 370-440 µg DNA/mg tissue
Mean Purity (A 260/280): 1.8

Nucleon™ BACC1: For 50 preps of 1 ml of whole blood or cultured cells (1 to 3×10⁶ cells/ml)

Nucleon™ BACC3: For 50 preps of 10 ml of whole blood (kit contains all reagents necessary for 50×10 ml blood extractions).