VWR® Maximum SF Shoe Covers, Fluid Impervious, Butterfly-Style
Supplier: VWR Collection
Certificates
About this item
VWR® Maximum SF shoe covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP), with breathable polyethylene film coating, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Non skid
- Fluid and particle impervious, sealed seams to repel water
- Superior friction coefficient to ensure maximum traction
- Maximum skid resistance on wet and smooth surfaces
- Clean, low linting fabric
Tested and manufactured in an ISO certified facility under stringent process controls. Lot controlled and validated through independent laboratory testing.
Length: 394 mm (U*), 432 mm (XL), 483 mm (XXL).
One size fits all.
Specifications
- Catalog No:
- 113-1380
- 113-1381
- 113-1367
- 113-1366
- 113-1379
- 113-1368
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory