Xylene cyanole FF ≥75% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:2650-17-1
:
Formula:C₂₅H₂₇N₂NaO₆S₂
MDL Number:MFCD00040657
:538.62 g/mol
Melting Pt:295 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
EINECS:220-167-5
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Light Green to Very Dark Green and Purple to Very Dark Purple and Black
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength in Water, C=0.01 G/L H2O:612 - 618 nm
- Extinction Coefficient:≥65000
- Carbon:≥41.2 %
- Nitrogen:≥3.8 %
- Purity (HPLC):≥96.0 %
- Solubility (Colour), C=1 mg/ml, H₂O:Dark Blue
Specification Test Results
Frequently Bought Together