Centrifugal devices, Microsep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
These centrifugal devices provide a precise, quick recovery of microlitre volumes of concentrate.
- High recoveries, typically greater than 90%
- Deadstop to prevent samples from spinning to dryness
- Fits centrifuges that accept standard 17×100 mm tubes
Applications: Concentrate dilute protein samples prior to electrophoresis. Exchange buffer and remove salt in samples. Remove proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies. Isolate low molecular weight compounds from fermentation broths for natural product screening. Recover biomolecules from cell culture supernatants or lysates. Clarify samples with gross particulate.
Quickly concentrates up to 20 ml of biological sample without valuable sample loss.
Specifications
- Catalog No:
- 516-0383
- 516-0382
- 516-0381
- 516-0378
- 516-0373
- 516-0374
- 516-0485
- 516-0377
- 516-0371
- 516-0370
- 516-9920
- 516-0376
- 516-0375
- 516-0372
- Type:
- Microsep™ Advance with Supor® membrane
- Microsep™ Advance with Supor® membrane
- Microsep™ Advance with Supor® membrane
- Microsep™ Advance with Supor® membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Microsep™ Advance with Omega™ membrane
- Filtration area:
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- 3,3 cm²
- Max. RCF:
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- 14000×g
- Dead volume:
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- 40 µl
- Ø×H:
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- 17×120 mm
- Sample volume:
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Max. 5000 µl
- Temperature:
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
Specifications
Frequently Bought Together