Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Overalls, SPP
VWR® Basic Cleanroom Overalls, SPP
VWR® Basic Cleanroom Overalls, SPP
Supplier:  VWR Collection
Certificates
VWR® Basic Cleanroom Overalls, SPP
7 Options Available Below
Product Details & Documents
VWR® Basic SPP overalls are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Tapered collar, inset sleeves, zip closure, elasticated cuffs
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms.
Tested and manufactured in an ISO certified facility also lot-controlled and validated through independent laboratory testing.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-1104
  • 113-1105
  • 113-1106
  • 113-1107
  • 113-1108
  • 113-1109
  • 113-1110
  • Garment application:
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
  • Cleanroom|General Purpose|Laboratory
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • Without hood
    White
    M
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 107,00 €
    107,00 €
    1 * 25 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1104
    Garment application
    Cleanroom|General Purpose|Laboratory
  • Without hood
    White
    L
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 107,00 €
    107,00 €
    1 * 25 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1105
    Garment application
    Cleanroom|General Purpose|Laboratory
  • Without hood
    White
    XL
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 107,00 €
    107,00 €
    1 * 25 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1106
    Garment application
    Cleanroom|General Purpose|Laboratory
  • Without hood
    White
    XXL
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1107
    Garment application
    Cleanroom|General Purpose|Laboratory
  • Without hood
    White
    3XL
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1108
    Garment application
    Cleanroom|General Purpose|Laboratory
  • 113-1109
    Without hood
    Without hood
    White
    4XL
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1109
    Garment application
    Cleanroom|General Purpose|Laboratory
  • 113-1110
    Without hood
    Without hood
    White
    5XL
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1110
    Garment application
    Cleanroom|General Purpose|Laboratory