Orders LinkContactUsLinkComponent
VWR® Basic Frocks, SPP
VWR® Basic Frocks, SPP
VWR® Basic Frocks, SPP
Supplier:  VWR Collection
Certificates
VWR® Basic Frocks, SPP
5 Options Available Below
Product Details & Documents
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Snap collar, raglan sleeves and snap closure
  • Elasticated cuffs, without pockets
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-1152
  • 113-1151
  • 113-1154
  • 113-1155
  • 113-1153
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Weight:
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    L
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 107,00 €
    107,00 €
    1 * 30 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1152
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory
    Weight
    48 g/m²
  • White
    M
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 107,00 €
    107,00 €
    1 * 30 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1151
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory
    Weight
    48 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 116,00 €
    116,00 €
    1 * 30 ST
       116,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1154
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory
    Weight
    48 g/m²
  • White
    3XL
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 116,00 €
    116,00 €
    1 * 30 ST
       116,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1155
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory
    Weight
    48 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 107,00 €
    107,00 €
    1 * 30 ST
       107,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1153
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory
    Weight
    48 g/m²