Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4864.
Supplier:  Avantor
CAS Number:  
Sodium hydroxide concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4864.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    60 mL
  • Seq:
    2385
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0099-0.01010

Specification Test Results

About this item

Synonyms: Caustic soda
 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548