Order Entry
Austria
ContactUsLinkComponent
 
Penicillinase (beta-Lactamase) Typ I (from Enterobacter cloacae)
 :  Merck
 
Penicillinase (beta-Lactamase) Typ I (from Enterobacter cloacae)
 :  Merck

 

  • CAS No.:
    9001-74-5
  • Source:
    Bacillus cereus
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Penicillinase (beta-Lactamase), Typ I

 

Frequently Bought Together