Order Entry
Austria
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Catalog # ROCH10621706001
Supplier:  ROCHE
CAS Number:  
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Catalog # ROCH10621706001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :833.35 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:220-163-3
CAS Number:2646-71-1
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263