Order Entry
Austria
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Catalog # ROCH10128040001
Supplier:  ROCHE
CAS Number:  
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Catalog # ROCH10128040001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :787.37 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
EINECS:246-129-8
CAS Number:24292-60-2
Formula:C₂₁H₂₆N₇Na₂O₁₇P₃
MDL Number:MFCD00065390