Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate 9% in aqueous solution Reag. Ph. Eur. 1033301
Disodium hydrogen phosphate dodecahydrate 9% in aqueous solution Reag. Ph. Eur. 1033301
Catalog # 87825.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Disodium hydrogen phosphate dodecahydrate 9% in aqueous solution Reag. Ph. Eur. 1033301
Catalog # 87825.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :358.14 g/mol
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10039-32-4
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181