Order Entry
Austria
ContactUsLinkComponent
Formaldehyde 35% in aqueous solution stabilised Reag. Ph. Eur. 1039101
Formaldehyde 35% in aqueous solution stabilised Reag. Ph. Eur. 1039101
Catalog # 87837.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Formaldehyde 35% in aqueous solution stabilised Reag. Ph. Eur. 1039101
Catalog # 87837.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
 :
 :30.03 g/mol
ADR:3 (8),III
 :Stabilised with methanol 9.0- 15.0 % Vol.
EINECS:200-001-8
UN:1198
 :GHS05
 :GHS08
 :GHS06
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274